missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethyl Ether, 98+%, Extra Pure

Product Code. 10105253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Metal drum
Unit Size:
10kg
1kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10105253 10 kg Metal drum 10kg
10713661 500 g Glass bottle 500g
10326940 1 kg Glass bottle 1kg
10090810 2.5 kg Metal drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10105253 Supplier Thermo Scientific Acros Supplier No. 117900100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-36-7 | C8H18O3 | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-36-7
MDL Number MFCD00009254
InChI Key RRQYJINTUHWNHW-UHFFFAOYSA-N
Synonym diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl
PubChem CID 8179
ChEBI CHEBI:44664
IUPAC Name 1-ethoxy-2-(2-ethoxyethoxy)ethane
SMILES CCOCCOCCOCC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -44.3°C
Density 0.9000g/mL
Boiling Point 180°C to 190°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Metal drum
Molecular Formula C8H18O3
Linear Formula (C2H5OCH2CH2)2O
Refractive Index 1.4105 to 1.4125
Quantity 10 kg
Beilstein 01, II, 519
Residue after Evaporation 0.05% max.
Merck Index 15, 3132
Specific Gravity 0.9
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in alcohol and other organic solvents,soluble in hydrocarbons,soluble in ether
Molecular Weight (g/mol) 162.23
Viscosity 1.4 mPa.s (20°C)
Formula Weight 162.23
Percent Purity 98+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethyl ether
Show More Show Less
Product Identifier
  • 2-Ethoxyethyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.