missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxyethyl Acetate, TRC
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
Specifications
Specifications
| Chemical Name or Material | Acetic acid-2-ethoxyethyl ester |
| CAS | 111-15-9 |
| Molecular Formula | C6 H12 O3 |
| Quantity | 25 mL |
| Synonym | Ethanol, 2-ethoxy-, acetate (6CI,7CI,8CI,9CI), 1-Acetoxy-2-ethoxyethane, 2-Ethoxyethanol acetate, 2-Ethoxyethyl acetate, 2-Ethyloxyethyl acetate, Cellosolve acetate, Ethyl cellosolve acetate, Ethyl glycol acetate, Ethylene glycol acetate monoethyl ether, Ethylene glycol ethyl ether acetate, Ethylene glycol monoethyl ether acetate, Ethylene glycol monoethyl ether monoacetate, Glycol monoethyl ether acetate, NSC 8658, O-Ethylglycol acetate, Oxitol acetate, Poly-Solv EE acetate, β-Ethoxyethyl acetate |
| InChI Formula | InChI=1S/C6H12O3/c1-3-8-4-5-9-6(2)7/h3-5H2,1-2H3 |
| SMILES | CCOCCOC(=O)C |
| IUPAC Name | 2-ethoxyethyl acetate |
| Molecular Weight (g/mol) | 132.16 |
| Formula Weight | 132.0786 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction