missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethyl Acetate, TRC

Product Code. 30374334
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 30374334

Brand: Toronto Research Chemicals TRCE6782605ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetic acid-2-ethoxyethyl ester
CAS 111-15-9
Molecular Formula C6 H12 O3
Quantity 5 mL
Synonym Ethanol, 2-ethoxy-, acetate (6CI,7CI,8CI,9CI), 1-Acetoxy-2-ethoxyethane, 2-Ethoxyethanol acetate, 2-Ethoxyethyl acetate, 2-Ethyloxyethyl acetate, Cellosolve acetate, Ethyl cellosolve acetate, Ethyl glycol acetate, Ethylene glycol acetate monoethyl ether, Ethylene glycol ethyl ether acetate, Ethylene glycol monoethyl ether acetate, Ethylene glycol monoethyl ether monoacetate, Glycol monoethyl ether acetate, NSC 8658, O-Ethylglycol acetate, Oxitol acetate, Poly-Solv EE acetate, β-Ethoxyethyl acetate
InChI Formula InChI=1S/C6H12O3/c1-3-8-4-5-9-6(2)7/h3-5H2,1-2H3
SMILES CCOCCOC(=O)C
IUPAC Name 2-ethoxyethyl acetate
Molecular Weight (g/mol) 132.16
Formula Weight 132.0786
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.