missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, for analysis

Product Code. 12357163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12357163 1 L 1 L
12367163 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12357163 Supplier Thermo Scientific Acros Supplier No. 445760010
  • ONLINE EXCLUSIVE PRICE
    £74.10
    £71.90 / 1 L
    Save £2.20
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-80-5 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002869
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
PubChem CID 8076
ChEBI CHEBI:46788
IUPAC Name 2-ethoxyethanol
SMILES CCOCCO

Specifications

Melting Point -70°C
Density 0.9300g/mL
Boiling Point 135.0°C
Flash Point 40°C
Infrared Spectrum Authentic
Linear Formula C2H5OCH2CH2OH
Refractive Index 1.4060 to 1.4100
Quantity 1 L
Residue after Evaporation 0.005% max.
Merck Index 15,3803
Specific Gravity 0.93
Solubility Information Solubility in water: complete solubility. Other solubilities: miscible with most organic solvents
Viscosity 2.08 mPa.s (20°C)
Formula Weight 90.12
Percent Purity ≥98.5% (GC)
Grade Analysis
Chemical Name or Material 2-Ethoxyethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Ethoxyethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.