missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, 99%, Thermo Scientific Chemicals

Product Code. 11473127
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473127 2500 mL 2500mL
11463127 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11473127 Supplier Thermo Scientific Alfa Aesar Supplier No. A16100.0F
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

A widely used solvent for nitrocelluloses and dyes2-Ethoxyethanol is widely used as a varnish remover and degreasing solution. It is a solvent used for nitrocellulose. It is used as a component for dyes, inks, cleaning agents, resins, paints, destaining solution for lipoproteins on polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A widely used solvent for nitrocelluloses and dyes2-Ethoxyethanol is widely used as a varnish remover and degreasing solution. It is a solvent used for nitrocellulose. It is used as a component for dyes, inks, cleaning agents, resins, paints, destaining solution for lipoproteins on polyacrylamide gels.

Solubility
Miscible with water, acetone, ethyl acetate, benzene, carbon tetrachloride, diethyl ether, methanol, hydrocarbons and ethanol.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents, aluminum, alkalies and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002869
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
PubChem CID 8076
ChEBI CHEBI:46788
IUPAC Name 2-ethoxyethan-1-ol
SMILES CCOCCO

Specifications

Melting Point -90°C
Density 0.931
Boiling Point 134°C to 135°C
Flash Point 44°C (111°F)
Odor Ether-like
Linear Formula CH3CH2OCH2CH2OH
Refractive Index 1.4085
Quantity 2500 mL
UN Number UN1171
Beilstein 1098271
Merck Index 14,3750
Solubility Information Miscible with water,acetone,ethyl acetate,benzene,carbon tetrachloride,diethyl ether,methanol,hydrocarbons and ethanol.
Formula Weight 90.12
Percent Purity 99%
Chemical Name or Material 2-Ethoxyethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Ethoxyethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Reproductive toxicity Category 1B
  • Acute toxicity Category 4
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.