missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxycyclohexanone, 98%

Product Code. 11426550
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426550 25 g 25g
11406550 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426550 Supplier Thermo Scientific Alfa Aesar Supplier No. H27613.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 33371-97-0 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33371-97-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00015377
InChI Key WBPWDGRYHFQTRC-QMMMGPOBSA-N
Synonym 2-ethoxycyclohexanone, ethoxycyclohexanone, 2-ethoxy-cyclohexanone, acmc-1ai48
PubChem CID 2733330
SMILES CCO[C@H]1CCCCC1=O

Specifications

Density 1.002
Boiling Point 85°C (15 mmHg)
Flash Point 54°C (129°F)
Refractive Index 1.451
Quantity 25 g
UN Number UN1224
IUPAC Name 2-ethoxycyclohexan-1-one
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material 2-Ethoxycyclohexanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.