missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 15457785
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457785 5 g 5g
15447785 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457785 Supplier Thermo Scientific Alfa Aesar Supplier No. H30941.06
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 269409-99-6 | Molecular Formula: C15H21BO4 | Molecular Weight: 276.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 269409-99-6
Molecular Formula C15H21BO4
Molecular Weight (g/mol) 276.139
MDL Number MFCD03453660
InChI Key RFBZWPFBCXBBJS-UHFFFAOYSA-N
Synonym ethyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid pinacol ester, ethyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid, pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester, ethyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-carboethoxyphenylboronic acid pinacol ester, 2-ethoxycarbonyl benzeneboronic acid pinacol ester, ethyl 2-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzoate
PubChem CID 2773962
IUPAC Name ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C(=O)OCC

Specifications

Melting Point 57°C to 62°C
Quantity 5 g
Formula Weight 276.14
Percent Purity 97%
Chemical Name or Material 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.