missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethoxycarbonyl)benzeneboronic acid, 98%

Product Code. 11415610
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415610 1 g 1g
11425610 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415610 Supplier Thermo Scientific Alfa Aesar Supplier No. H25867.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 380430-53-5 | C9H11BO4 | 193.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380430-53-5
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02179453
InChI Key QZKVVOXAEBCLPZ-UHFFFAOYSA-N
Synonym 2-ethoxycarbonyl phenyl boronic acid, 2-ethoxycarbonylbenzeneboronic acid, 2-ethoxycarbonyl phenylboronic acid, 2-ethoxycarbonylphenyl boronic acid, o-ethoxycarbonylphenylboronic acid, 2-carbethoxyphenylboronic acid, ethyl 2-dihydroxyboranyl benzoate, ethyl 2-boronobenzoate, 2-ethoxycarbonyl benzeneboronic acid
PubChem CID 2773405
IUPAC Name (2-ethoxycarbonylphenyl)boronic acid
SMILES CCOC(=O)C1=CC=CC=C1B(O)O

Specifications

Melting Point 130°C to 133°C
Quantity 1 g
Formula Weight 193.99
Percent Purity 98%
Chemical Name or Material 2-(Ethoxycarbonyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.