missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzonitrile, 98%, Thermo Scientific™

Product Code. 10037899 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10037899 5 g 5g
10453160 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10037899 Supplier Thermo Scientific Acros Supplier No. 435340050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6609-57-0
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00001789
InChI Key DXTLCLWOCYLDHL-UHFFFAOYSA-N
Synonym o-ethoxybenzonitrile, benzonitrile, ethoxy, 2-ethoxybenzenecarbonitrile, o-ethoxybenzontrle, acmc-20anwu, 2-ethoxy benzonitrile, 2-ethoxy-benzonitrile, pubchem4773, benzonitrile, 2-ethoxy, 2-ethoxybenzonitrile
PubChem CID 576922
IUPAC Name 2-ethoxybenzonitrile
SMILES CCOC1=CC=CC=C1C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5°C
Color Colorless to Yellow
Boiling Point 45°C (0.5 Torr)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 147.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Ethoxybenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2-Ethoxybenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.