missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzoic acid, 98%

Product Code. 11437816
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437816 500 g 500g
11417816 25 g 25g
11427816 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437816 Supplier Thermo Scientific Alfa Aesar Supplier No. A14758.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 134-11-2 | C9H10O3 | 166.18 g/mol

2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-11-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00002438
InChI Key XDZMPRGFOOFSBL-UHFFFAOYSA-N
Synonym o-ethoxybenzoic acid, benzoic acid, 2-ethoxy, benzoic acid, o-ethoxy, o-ethylsalicylic acid, benzoic acid, ethoxy, 2'-carboxyphenetole, 2-ethoxy benzoic acid, 2-ethoxy-benzoic acid, pubchem20122, ortho-ethoxybenzoic acid
PubChem CID 67252
IUPAC Name 2-ethoxybenzoic acid
SMILES CCOC1=CC=CC=C1C(O)=O

Specifications

Melting Point 19°C to 20°C
Density 1.105
Boiling Point 174°C to 175°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.541
Quantity 500 g
Beilstein 2691039
Solubility Information Soluble in water.
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2-Ethoxybenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.