missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzamidine Hydrochloride, TRC

Product Code. 30388106
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388106 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388106 Supplier Toronto Research Chemicals Supplier No. TRCE89056025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethoxybenzamidine Hydrochloride
CAS 18637-00-8
Molecular Formula C9H13ClN2O
Quantity 25 g
Synonym 2-Ethoxybenzenecarboximidamide Hydrochloride, o-Ethoxybenzamidine Monohydrochloride, 2-Ethoxybenzenecarboximidamide Monohydrochloride
InChI Formula InChI=1S/C9H12N2O.ClH/c1-2-12-8-6-4-3-5-7(8)9(10)11;/h3-6H,2H2,1H3,(H3,10,11);1H
SMILES N=C(N)C1=CC=CC=C1OCC.Cl
Molecular Weight (g/mol) 200.67
Formula Weight 200.0716
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.