missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzamide, MedChemExpress

Product Code. 16402349
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402349 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402349 Supplier Medchem Express Supplier No. HYB1428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 2-Ethoxybenzamide is widely used as an antipyretic anodyne.

2-Ethoxybenzamide is widely used as an antipyretic anodyne.

Specifications

Chemical Name or Material 2-Ethoxybenzamide
CAS 938-73-8
Color White
Purity Grade Notes Research
Molecular Formula C9H11NO2
Quantity 1 g
Synonym Ethenzamide
Solubility Information DMSO : ≥ 47 mg/mL (284.52 mM)
SMILES O=C(N)C1=CC=CC=C1OCC
Molecular Weight (g/mol) 165.19
Formula Weight 165.19
Percent Purity 99.78%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.