missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzamide, 97%

Product Code. 11467136
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467136 100 g 100g
11477136 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467136 Supplier Thermo Scientific Alfa Aesar Supplier No. B25455.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 938-73-8 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-73-8
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00007977
InChI Key SBNKFTQSBPKMBZ-UHFFFAOYSA-N
Synonym ethenzamide, benzamide, 2-ethoxy, o-ethoxybenzamide, ethbenzamide, etenzamide, ethenzamid, etamide, ethosalicyl, etosalicil, etosalicyl
PubChem CID 3282
IUPAC Name 2-ethoxybenzamide
SMILES CCOC1=CC=CC=C1C(N)=O

Specifications

Melting Point 131°C to 134°C
Quantity 100 g
Merck Index 14,3731
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material 2-Ethoxybenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.