missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzamide, 97%

Product Code. 10060800 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10060800 100 g Plastic bottle 100g
10468640 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10060800 Supplier Thermo Scientific Acros Supplier No. 117871000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 938-73-8 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-73-8
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00007977
InChI Key SBNKFTQSBPKMBZ-UHFFFAOYSA-N
Synonym ethenzamide, benzamide, 2-ethoxy, o-ethoxybenzamide, ethbenzamide, etenzamide, ethenzamid, etamide, ethosalicyl, etosalicil, etosalicyl
PubChem CID 3282
IUPAC Name 2-ethoxybenzamide
SMILES CCOC1=CC=CC=C1C(N)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 129.0°C to 134.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula C2H5OC6H4CONH2
Quantity 100 g
Beilstein 10, 93
Merck Index 15, 3786
Solubility Information Solubility in water: slightly soluble in water.
Formula Weight 165.19
Percent Purity 97%
Physical Form Granular Crystalline Powder
Chemical Name or Material 2-Ethoxybenzamide, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.