missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzaldehyde, 97+%

Product Code. 11427287
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427287 50 g 50g
11349629 100 g 100g
11437287 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427287 Supplier Thermo Scientific Alfa Aesar Supplier No. A15035.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 613-69-4 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-69-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00003316
InChI Key DUVJMSPTZMCSTQ-UHFFFAOYSA-N
Synonym benzaldehyde, 2-ethoxy, o-ethoxybenzaldehyde, 2-ethoxy benzaldehyde, benzaldehyde, o-ethoxy, 2-ethoxy-benzaldehyde, benzaldehyde, ethoxy, ethoxy benzaldehyde, pubchem9726, ortho-ethoxybenzaldehyde, acmc-209ms1
PubChem CID 11950
IUPAC Name 2-ethoxybenzaldehyde
SMILES CCOC1=CC=CC=C1C=O

Specifications

Density 1.077
Boiling Point 249°C
Flash Point 91°C (195°F)
Refractive Index 1.543
Quantity 50 g
Beilstein 1937270
Sensitivity Air Sensitive
Formula Weight 150.18
Percent Purity ≥97%
Chemical Name or Material 2-Ethoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.