missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyaniline, TRC

Product Code. 30132754
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132754 10 g 10g
30132967 5 g 5g
18020920 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132754 Supplier Toronto Research Chemicals Supplier No. TRCE20046110G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-70-2
Molecular Formula C8 H11 N O
Molecular Weight (g/mol) 137.18
Synonym o-Phenetidine (7CI,8CI), 2-Ethoxybenzenamine, 2-Aminophenetole, 2-Aminophenyl ethyl ether, 2-Ethoxyaniline, 2-Ethoxyphenylamine, NSC 9818, o-Ethoxyaniline, o-Ethoxylaniline, o-Phenetidine, 2-Phenetidine, 2-Ethoxybenzenamine, 2-Aminophenetole, 2-Aminophenyl Ethyl Ether, 2-Ethoxyphenylamine, o-Ethoxyaniline, o-Phenetidine (7CI,8CI), 2-Ethoxyaniline, NSC 9818, 2-Phenetidine
IUPAC Name 2-ethoxyaniline
SMILES CCOc1ccccc1N

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H11NO/c1-2-10-8-6-4-3-5-7(8)9/h3-6H,2,9H2,1H3
Formula Weight 137.0841
Chemical Name or Material 2-Ethoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.