missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyacetyl Chloride, TRC

Product Code. 30382036
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382036 5 g -
30379439 10 g -
30371570 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382036 Supplier Toronto Research Chemicals Supplier No. TRCE8909305G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 14077-58-8
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.55
Synonym alpha-Ethoxyacetyl Chloride, Ethoxyacetic Acid Chloride
IUPAC Name 2-ethoxyacetyl chloride
SMILES CCOCC(=O)Cl

Specifikationer

Quantity 5 g
InChI Formula InChI=1S/C4H7ClO2/c1-2-7-3-4(5)6/h2-3H2,1H3
Formula Weight 122.0135
Chemical Name or Material 2-Ethoxyacetyl Chloride
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.