missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyacetyl chloride, 97%, stab. with 0.3% Magnesium oxide

Product Code. 15488945
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488945 100 g 100g
15468945 5 g 5g
15478945 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15488945 Supplier Thermo Scientific Alfa Aesar Supplier No. H34443.22
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14077-58-8 | C4H7ClO2 | 122.548 g/mol

It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.

Solubility
Insoluble in water.

Notes
Reacts with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14077-58-8
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD11170566
InChI Key ZPMWWAIBJJFPPQ-UHFFFAOYSA-N
PubChem CID 84189
IUPAC Name 2-ethoxyacetyl chloride
SMILES CCOCC(=O)Cl

Specifications

Linear Formula CH3CH2OCH2COCl
Quantity 100 g
UN Number UN2920
Synonym ethoxyacetyl chloride, ethoxyacetylchloride, ethoxy-acetyl chloride, acetyl chloride,2-ethoxy, 2-ethoxyacetylchloride, ethoxy acetyl chloride, ethoxyacetic acid chloride, 2-ethoxy-acetyl chloride, acetyl chloride, 2-ethoxy
Solubility Information Insoluble in water.
Formula Weight 122.55
Percent Purity 97%
Chemical Name or Material 2-Ethoxyacetyl chloride, stabilized with 0.3% Magnesium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.