Learn More
2-Ethoxyacetyl chloride, 97%, stab. with 0.3% Magnesium oxide
Description
It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.
Solubility
Insoluble in water.
Notes
Reacts with strong oxidizing agents. Store at room temperature.
Specifications
Specifications
| Linear Formula | CH3CH2OCH2COCl |
| Quantity | 5 g |
| UN Number | UN2920 |
| Synonym | ethoxyacetyl chloride, ethoxyacetylchloride, ethoxy-acetyl chloride, acetyl chloride,2-ethoxy, 2-ethoxyacetylchloride, ethoxy acetyl chloride, ethoxyacetic acid chloride, 2-ethoxy-acetyl chloride, acetyl chloride, 2-ethoxy |
| Solubility Information | Insoluble in water. |
| Formula Weight | 122.55 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Ethoxyacetyl chloride, stabilized with 0.3% Magnesium oxide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.