missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-5-iodonicotinic acid, 97%, Thermo Scientific™

Product Code. 10321710
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10321710 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10321710 Supplier Thermo Scientific Maybridge Supplier No. MAY00289DA
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 335078-07-4
Assay Percent Range 97%
Molecular Formula C8H8INO3
Quantity 1 g
Synonym 2-ethoxy-5-iodonicotinic acid, 2-ethoxy-5-iodo-pyridine-3-carboxylic acid, 3-carboxy-2-ethoxy-5-iodopyridine
InChI Key DHVUWACWJRDVBQ-UHFFFAOYSA-N
SMILES CCOC1=NC=C(C=C1C(=O)O)I
IUPAC Name 2-ethoxy-5-iodopyridine-3-carboxylic acid
Molecular Weight (g/mol) 293.06
PubChem CID 22648229
Formula Weight 293.06
Chemical Name or Material 2-Ethoxy-5-iodonicotinic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.