missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-2-oxoethoxy Digoxigenin, TRC

Product Code. 30380810
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380810 100 mg 100mg
30384188 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380810 Supplier Toronto Research Chemicals Supplier No. TRCE677980100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethoxy-2-oxoethoxy Digoxigenin
Analyte or Component Names 2-Ethoxy-2-oxoethoxy Digoxigenin
Shipping Condition Room Temperature
Molecular Formula C27 H40 O7
Synonym (3beta,5beta,12beta)-12-Hydroxy-3-(2-ethoxy-2-oxoethoxy)-14-hydroxycard-20(22)-enolide
InChI Formula InChI=1S/C27H40O7/c1-4-32-24(30)15-33-18-7-9-25(2)17(12-18)5-6-20-21(25)13-22(28)26(3)19(8-10-27(20,26)31)16-11-23(29)34-14-16/h11,17-22,28,31H,4-10,12-15H2,1-3H3/t17-,18+,19-,20-,21+,22-,25+,26+,27+/m1/s1
Solution Type Neat
SMILES CCOC(=O)CO[C@H]1CC[C@@]2(C)[C@H](CC[C@@H]3[C@@H]2C[C@@H](O)[C@]4(C)[C@H](CC[C@]34O)C5=CC(=O)OC5)C1
IUPAC Name ethyl 2-[[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-12,14-dihydroxy-10,13-dimethyl-17-(5-oxo-2H-furan-3-yl)-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-yl]oxy]acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 476.602
Quantity 100 mg
Formula Weight 476.277
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.