missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-1-naphthoyl chloride, 97%, Thermo Scientific™

Product Code. 11431007
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431007 1 g 1g
11441007 5 g 5g
11451007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431007 Supplier Thermo Scientific Alfa Aesar Supplier No. B23399.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55150-29-3
Molecular Formula C13H11ClO2
Molecular Weight (g/mol) 234.68
MDL Number MFCD00046426
InChI Key ZRVFFIXOCFUHDA-UHFFFAOYSA-N
Synonym 2-ethoxy-1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 2-ethoxy, 1-naphthalenecarbonylchloride, 2-ethoxy, acmc-209ll8, 2-ethoxy naphthoyl chloride, 2-ethoxy-1-naphthoylchloride, 2-ethoxy-1-naphthalenecarbonyl chloride, 2-ethoxy-1-naphthalenecarboxyl chloride, 2-ethoxy-naphthalene-1-carbonyl chloride
PubChem CID 108679
IUPAC Name 2-ethoxynaphthalene-1-carbonyl chloride
SMILES CCOC1=CC=C2C=CC=CC2=C1C(Cl)=O

Specifications

Melting Point 76°C to 80°C
Quantity 1 g
UN Number UN3261
Beilstein 2844104
Sensitivity Moisture sensitive
Formula Weight 234.68
Percent Purity 97%
Chemical Name or Material 2-Ethoxy-1-naphthoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.