missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, 99%

Product Code. p-7021405
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11454734

Brand: Thermo Scientific Alfa Aesar A13724.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16357-59-8 | C14H17NO3 | 247.29 g/mol

2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is an irreversible membrane-bound receptor antagonist. It is used in synthetic chemistry, discovery, process R&D. It also acts as coupling reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline is an irreversible membrane-bound receptor antagonist. It is used in synthetic chemistry, discovery, process R&D. It also acts as coupling reagents.

Solubility
Soluble in 95% Ethanol (50 mg/ml). Insoluble in water.

Notes
Moisture Sensitive. Store at 4°C. Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16357-59-8
Molecular Formula C14H17NO3
Molecular Weight (g/mol) 247.29
MDL Number MFCD00006703
InChI Key GKQLYSROISKDLL-UHFFFAOYNA-N
Synonym eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate
PubChem CID 27833
IUPAC Name ethyl 2-ethoxy-2H-quinoline-1-carboxylate
SMILES CCOC1C=CC2=CC=CC=C2N1C(=O)OCC

Specifications

Melting Point 63°C to 66°C
Boiling Point 125°C to 128°C (0.1 mmHg)
Quantity 10 g
Beilstein 533048
Sensitivity Moisture sensitive
Merck Index 14,3518
Solubility Information Soluble in 95% Ethanol (50mg/ml). Insoluble in water.
Formula Weight 247.29
Percent Purity 99%
Chemical Name or Material 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.