missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-1-[[2'-[2-(trityl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-4-carboxylic Acid Methyl Ester-d4 (Candesartan Impurity), TRC

Product Code. 30381855
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381855 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30381855 Supplier Toronto Research Chemicals Supplier No. TRCE892996100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethoxy-1-((2'-(2-(trityl)-2H-tetrazol-5-yl)(1,1'-biphenyl)-4-yl)methyl)-1H-benzimidazole-4-carboxylic Acid Methyl Ester-d4 (Candesartan Impurity)
CAS 1793997-59-7
Molecular Formula C44H32D4N6O3
Quantity 100 mg
Synonym 2-Ethoxy-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-4-carboxylic Acid Methyl Ester-d4
InChI Formula InChI=1S/C44H36N6O3/c1-3-53-43-45-40-38(42(51)52-2)24-15-25-39(40)49(43)30-31-26-28-32(29-27-31)36-22-13-14-23-37(36)41-46-47-48-50(41)44(33-16-7-4-8-17-33,34-18-9-5-10-19-34)35-20-11-6-12-21-35/h4-29H,3,30H2,1-2H3/i26D,27D,28D,29D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1Cn2c(OCC)nc3c(cccc23)C(=O)OC)c4ccccc4c5nnnn5C(c6ccccc6)(c7ccccc7)c8ccccc8
IUPAC Name methyl 2-ethoxy-1-[[2,3,5,6-tetradeuterio-4-[2-(1-trityltetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
Molecular Weight (g/mol) 700.82
Formula Weight 700.31
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.