missing translation for 'onlineSavingsMsg'
Learn More

2-Ethenyl-2-methyl-5-prop-1-en-2-yloxolane, TRC

Product Code. 30331781
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331781 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331781 Supplier Toronto Research Chemicals Supplier No. TRCB1253132.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethenyl-2-methyl-5-prop-1-en-2-yloxolane
CAS 13679-86-2
Molecular Formula C10H16O
Quantity 2.5 g
InChI Formula InChI=1S/C10H16O/c1-5-10(4)7-6-9(11-10)8(2)3/h5,9H,1-2,6-7H2,3-4H3
SMILES CC(=C)C1CCC(C)(O1)C=C
IUPAC Name 2-ethenyl-2-methyl-5-prop-1-en-2-yloxolane
Molecular Weight (g/mol) 152.23
Formula Weight 152.1201
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.