Learn More
JULABO™ 2-Elbow Fittings
Product Code. 15294293
missing translation for 'orderingAttributeHoverText'
Size:
M24x1.5 female/male
M30x1.5 female/male
M38 x 1.5 female/male
missing translation for 'unitSize'
Pack of 2
Product Code. 15294293
missing translation for 'mfr': JULABO™ 8890122
Only null left
Add to basket
Add to basket
For use with PRESTO™ Temperature Control System/Process System. Metal construction with 90° angle.
  • For use with PRESTO™ Temperature Control System/Proecess System
  • 2 Elbow fittings, 90°
  • Metal construction
  • Choose:
    • M24x1.5 female/male
    • M30x1.5 female/male
    • M38x1.5 female/male
Specifications
For Use With (Equipment) JULABO™ PRESTO™
Type 2-Elbow Fittings
Size M38 x 1.5 female/male