missing translation for 'onlineSavingsMsg'
Learn More

2-Dodecylisoquinolinium Bromide, TRC

Product Code. 30370556
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370556 1 g 1g
30385972 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30370556 Supplier Toronto Research Chemicals Supplier No. TRCD5255401G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Dodecylisoquinolinium Bromide
CAS 93-23-2
Molecular Formula C21H32BrN
Quantity 1 g
Synonym 2-Dodecylisoquinolinium bromide (6CI,7CI), Isoquinolinium, 2-dodecyl-, bromide (8CI,9CI), 2-Dodecylisoquinolin-2-ium bromide, Decelerine, Dodecylisoquinolinium bromide, Isothan, Isothan Q 15, Isothan Q-90, Laurylisoquinolinium bromide, N-Laurylisoquinolinium bromide, n-Dodecylisoquinolinium bromide, Isoquinolinium, 2-dodecyl-, bromide (1:1), Decelerine, Dodecylisoquinolinium Bromide, Isothan, Isothan Q 15, Isothan Q-90, Laurylisoquinolinium Bromide, N-Laurylisoquinolinium Bromide, n-Dodecylisoquinolinium Bromide
InChI Formula InChI=1S/C21H32N.BrH/c1-2-3-4-5-6-7-8-9-10-13-17-22-18-16-20-14-11-12-15-21(20)19-22;/h11-12,14-16,18-19H,2-10,13,17H2,1H3;1H/q+1;/p-1
SMILES [Br-].CCCCCCCCCCCC[n+]1ccc2ccccc2c1
IUPAC Name 2-dodecylisoquinolin-2-ium;bromide
Molecular Weight (g/mol) 378.39
Formula Weight 377.1718
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.