missing translation for 'onlineSavingsMsg'
Learn More

2-Dodecanone, 95%

Product Code. 11483118
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483118 50 g 50g
11473118 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483118 Supplier Thermo Scientific Alfa Aesar Supplier No. A18862.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6175-49-1 | C12H24O | 184.323 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6175-49-1
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.323
MDL Number MFCD00015064
InChI Key LSKONYYRONEBKA-UHFFFAOYSA-N
Synonym 2-dodecanone, methyl decyl ketone, decyl methyl ketone, unii-p5cn8ysv3p, dodecanone-2, p5cn8ysv3p, n-decyl methyl ketone, dodecanone, acmc-1b2ve, 2-dodecanone gc
PubChem CID 22556
IUPAC Name dodecan-2-one
SMILES CCCCCCCCCCC(=O)C

Specifications

Melting Point 17°C to 20°C
Density 0.82
Boiling Point 247°C
Flash Point 110°C (230°F)
Refractive Index 1.433
Quantity 50 g
Beilstein 1703135
Formula Weight 184.32
Percent Purity ≥98%
Chemical Name or Material 2-Dodecanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.