missing translation for 'onlineSavingsMsg'
Learn More

2-Dodecanone, 95%

Product Code. 10096253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10096253 100 mL Glass bottle 100mL
10183431 5 mL Glass bottle 5mL
10632862 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10096253 Supplier Thermo Scientific Acros Supplier No. 330291000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6175-49-1 | C12H24O | 184.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6175-49-1
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.32
MDL Number MFCD00015064
InChI Key LSKONYYRONEBKA-UHFFFAOYSA-N
Synonym 2-dodecanone, methyl decyl ketone, decyl methyl ketone, unii-p5cn8ysv3p, dodecanone-2, p5cn8ysv3p, n-decyl methyl ketone, dodecanone, acmc-1b2ve, 2-dodecanone gc
PubChem CID 22556
IUPAC Name dodecan-2-one
SMILES CCCCCCCCCCC(=O)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 17°C to 20°C
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 247°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4320 to 1.4340 (20°C, 589nm)
Quantity 100 mL
Specific Gravity 0.82
Formula Weight 184.32
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Dodecanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.