missing translation for 'onlineSavingsMsg'
Learn More

2-Docosahexaenoin-1-oleoyl 3-Phosphocholine, TRC

Product Code. 30370333
Change view
Click to view available options
Quantity:
5 mg
50 mg
Product Code. Quantity
30370333 5 mg
30369903 50 mg
2 options
This item is not returnable. View return policy

Product Code. 30370333

Brand: Toronto Research Chemicals TRCD4944805MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99296-82-9
InChI Formula InChI=1S/C48H82NO8P/c1-6-8-10-12-14-16-18-20-22-23-24-25-27-29-31-33-35-37-39-41-48(51)57-46(45-56-58(52,53)55-43-42-49(3,4)5)44-54-47(50)40-38-36-34-32-30-28-26-21-19-17-15-13-11-9-7-2/h8,10,14,16,20-22,24-26,29,31,35,37,46H,6-7,9,11-13,15,17-19,23,27-28,30,32-34,36,38-45H2,1-5H3/b10-8-,16-14-,22-20-,25-24-,26-21-,31-29-,37-35-/t46-/m1/s1
Molecular Formula C48H82NO8P
Molecular Weight (g/mol) 832.14
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CC
Synonym [R-(all-Z)]-4-Hydroxy-N,N,N-trimethyl-9-oxo-7-[[[(9Z)-1-oxo-9- octadecen-1-yl]oxy]methyl]-3,5,8-trioxa-4-phosphatriaconta-12,15,18,21,24,27-hexaen-1-aminium Inner Salt 4-Oxide
Chemical Name or Material 2-Docosahexaenoin-1-oleoyl 3-Phosphocholine

Specifications

Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.