missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Diphenylphosphinobiphenyl, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 2-(Diphenylphosphino)biphenyl |
| CAS | 13885-09-1 |
| Molecular Formula | C24H19P |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C24H19P/c1-4-12-20(13-5-1)23-18-10-11-19-24(23)25(21-14-6-2-7-15-21)22-16-8-3-9-17-22/h1-19H |
| SMILES | c1ccc(cc1)P(c2ccccc2)c3ccccc3c4ccccc4 |
| IUPAC Name | diphenyl-(2-phenylphenyl)phosphane |
| Molecular Weight (g/mol) | 338.38 |
| Formula Weight | 338.1224 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction