missing translation for 'onlineSavingsMsg'
Learn More

2-[(Diphenylmethyl)thio]acetic Acid, TRC

Product Code. 30380977
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380977 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380977 Supplier Toronto Research Chemicals Supplier No. TRCD491830500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 63547-22-8
Molecular Formula C15 H14 O2 S
Quantity 500 mg
InChI Formula InChI=1S/C15H14O2S/c16-14(17)11-18-15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H,16,17)
SMILES OC(=O)CSC(c1ccccc1)c2ccccc2
IUPAC Name 2-benzhydrylsulfanylacetic acid
Molecular Weight (g/mol) 258.34
Formula Weight 258.0715
Chemical Name or Material 2-[(Diphenylmethyl)thio]acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.