missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylcarbamoyl)benzeneboronic acid, 95%

Product Code. 15479497
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479497 250 mg 250mg
15469497 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479497 Supplier Thermo Scientific Alfa Aesar Supplier No. H52765.MD
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 874219-16-6 | C9H12BNO3 | 193.009 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874219-16-6
Molecular Formula C9H12BNO3
Molecular Weight (g/mol) 193.009
MDL Number MFCD03425959
InChI Key NZIOVLXULSCCSG-UHFFFAOYSA-N
Synonym 2-dimethylcarbamoyl phenyl boronic acid, 2-n,n-dimethylaminocarbonyl phenylboronic acid, 2-dimethylcarbamoyl phenylboronic acid, 2-dimethylaminocarbonyl benzeneboronic acid, boronic acid,b-2-dimethylamino carbonyl phenyl, 2-dimethylcarbamoyl benzeneboronic acid, acmc-209qkh, 2-n,n-dimethylaminocarbonyl benzeneboronic acid, 2-dimethylcarbamoyl phenyl boronicacid
PubChem CID 44119823
IUPAC Name [2-(dimethylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=CC=C1C(=O)N(C)C)(O)O

Specifications

Melting Point 108°C to 110°C
Quantity 250 mg
Formula Weight 193.01
Percent Purity 95%
Chemical Name or Material 2-(Dimethylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.