missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98%

Product Code. 10737972 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10737972 1 g Glass bottle 1g
10449412 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10737972 Supplier Thermo Scientific Acros Supplier No. 335620010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42036-65-7 | C9H17NO·HCl | 191.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42036-65-7
Molecular Formula C9H17NO·HCl
Molecular Weight (g/mol) 191.7
MDL Number MFCD00102580
InChI Key CLVHTSWMNNSUSH-UHFFFAOYSA-N
Synonym 2-dimethylamino methyl cyclohexanone hydrochloride, 2-dimethylaminomethyl-1-cyclohexanone hydrochloride, dimethyl 2-oxocyclohexyl methyl ammonium chloride, 2-dimethylamino methyl cyclohexan-1-one hydrochloride, 2-dimethylaminomethyl-cyclohexanone, 2-dimethylamino methyl cyclohexan-1-one, chloride, tramadol related compound b, 2-dimethylaminomethyl-cyclohexanone hydrochloride, tramadol related compound b usp
PubChem CID 359482
IUPAC Name 2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride
SMILES CN(C)CC1CCCCC1=O.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 147°C to 151°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol,ethanol
Formula Weight 191.7
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.