missing translation for 'onlineSavingsMsg'
Learn More

(2-Dimethylaminoethyl)triphenylphosphonium bromide, 98%

Product Code. 11344364
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344364 10 g 10g
11354364 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344364 Supplier Thermo Scientific Alfa Aesar Supplier No. L00917.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 21331-80-6 | C22H25BrNP | 414.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21331-80-6
Molecular Formula C22H25BrNP
Molecular Weight (g/mol) 414.33
MDL Number MFCD00011794
InChI Key YIIHEMGEQQWSMA-UHFFFAOYSA-M
Synonym 2-dimethylaminoethyl triphenylphosphonium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphonium bromide, 2-dimethylaminoethyl triphenylphosphoniumbromide, 2-dimethylamino ethyl triphenylphosphanium bromide, acmc-1ci7c, triphenyldimethylaminoethylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphoniumbromide
PubChem CID 2724136
IUPAC Name 2-(dimethylamino)ethyl-triphenylphosphanium;bromide
SMILES [Br-].CN(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∽200°C (decomposition)
Quantity 10 g
Beilstein 4072092
Sensitivity Hygroscopic
Formula Weight 414.33
Percent Purity 98%
Chemical Name or Material (2-Dimethylaminoethyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.