missing translation for 'onlineSavingsMsg'
Learn More

(2-Dimethylaminoethyl)triphenylphosphonium bromide, 97%, Thermo Scientific™

Product Code. 10194070 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10194070 50 g Glass bottle 50g
10782431 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10194070 Supplier Thermo Scientific Acros Supplier No. 201410500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21331-80-6
Molecular Formula C22H25BrNP
Molecular Weight (g/mol) 414.33
MDL Number MFCD00011794
InChI Key YIIHEMGEQQWSMA-UHFFFAOYSA-M
Synonym 2-dimethylaminoethyl triphenylphosphonium bromide, 2-dimethylamino ethyl triphenylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphonium bromide, 2-dimethylaminoethyl triphenylphosphoniumbromide, 2-dimethylamino ethyl triphenylphosphanium bromide, acmc-1ci7c, triphenyldimethylaminoethylphosphonium bromide, 2-dimethylaminoethyltriphenylphosphoniumbromide
PubChem CID 2724136
SMILES [Br-].CN(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 201.0°C
Color Brown-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)2NCH2CH2P(C6H5)3Br
Quantity 50 g
Fieser 08,184; 09,291; 13,174; 15,198; 16,205
IUPAC Name [2-(dimethylamino)ethyl]triphenylphosphanium bromide
Formula Weight 414.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (2-Dimethylaminoethyl)triphenylphosphonium bromide, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.