missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoethanol, TRC

Product Code. 30383694
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383694 50 g
30370638 10 g
30379625 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383694 Supplier Toronto Research Chemicals Supplier No. TRCD47117050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-01-0
Molecular Formula C4 H11 N O
Molecular Weight (g/mol) 89.14
Synonym 2-(Dimethylamino)ethanol, (2-Hydroxyethyl)dimethylamine, (Dimethylamino)ethanol, (N,N-Dimethylamino)ethanol, 2-(Dimethylamino)ethanol, 2-(Dimethylamino)ethyl alcohol, 2-(N,N-Dimethylamino)ethanol, Amietol M 21, Aminoalcohol 2mabs, Bimanol, DMAE, DMEA, Dabco DMEA, Deanol, DeuAdd MA 95, Dimethol, Dimethyl(2-hydroxyethyl)amine, Dimethyl(hydroxyethyl)amine, Dimethylethanolamine, Dimethylmonoethanolamine, Jeffcat DMEA, Kalpur P, Lupragen N 101, MA 95, N,N-Dimethyl(2-hydroxyethyl)amine, N,N-Dimethyl-2-aminoethanol, N,N-Dimethyl-2-ethanolamine, N,N-Dimethyl-N-(β-hydroxyethyl)amine, N,N-Dimethyl-β-hydroxyethylamine, N,N-Dimethylethanolamine, N-(2-Hydroxyethyl)-N,N-dimethylamine, N-(2-Hydroxyethyl)dimethylamine, NSC 2652, Niax DMEA, Norcholine, PC CAT DMEA, Propamine A, Rexolin, T 80, Tegoamin DMEA, Texacat DME, Texacat DMEA, Thancat DME, β-(Dimethylamino)ethanol, β-Dimethylaminoethyl alcohol, β-Hydroxyethyldimethylamine
IUPAC Name 2-(dimethylamino)ethanol
SMILES CN(C)CCO

Specifications

Quantity 50 g
InChI Formula InChI=1S/C4H11NO/c1-5(2)3-4-6/h6H,3-4H2,1-2H3
Formula Weight 89.0841
Percent Purity >95%
Chemical Name or Material N,N-Dimethylethanolamine
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.