missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoethanol Bitartrate, TRC

Product Code. 30386280
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386280 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386280 Supplier Toronto Research Chemicals Supplier No. TRCD471440100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Dimethylaminoethanol Bitartrate
CAS 5988-51-2
Molecular Formula C8H17NO7
Quantity 100 g
Synonym Deanol Bitartrate, Dimethylaminoethanol Bitartrate, Liparon, N,N-Dimethylethanolamine-tartaric Acid Salt, (2R,3R)-2,3-Dihydroxybutanedioate 2-(Dimethylamino)-ethanol, [R-(R*,R*)]-2,3-dihydroxybutanedioate 2-(Dimethylamino)-ethanol, 2-Dimethylaminoethanol Bitartrate, 2-Dimethylaminoethanol Tartrate
InChI Formula InChI=1S/C4H11NO.C4H6O6/c1-5(2)3-4-6;5-1(3(7)8)2(6)4(9)10/h6H,3-4H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1
SMILES CN(C)CCO.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;2-(dimethylamino)ethanol
Molecular Weight (g/mol) 239.22
Formula Weight 239.1005
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.