missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)pyridine, 97%

Product Code. p-7043751
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11327735

Brand: Thermo Scientific Alfa Aesar L02513.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5683-33-0 | C7H10N2 | 122.17 g/mol

2-(Dimethylamino)pyridine in toluene at room temperature in the presence of 1.1 equivalent of methyl trifluoromethanesulfonate forms (2-pyridyl)-trimethylammonium trifluoromethanesulfonate salt. It is used as a model substrate in the preparation of chelate carbene via cyclometalation, H2 loss and reversible α-elimination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Dimethylamino)pyridine in toluene at room temperature in the presence of 1.1 equivalent of methyl trifluoromethanesulfonate forms (2-pyridyl)-trimethylammonium trifluoromethanesulfonate salt. It is used as a model substrate in the preparation of chelate carbene via cyclometalation, H2 loss and reversible α-elimination.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5683-33-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00006261
InChI Key PSHKMPUSSFXUIA-UHFFFAOYSA-N
Synonym 2-dimethylaminopyridine, 2-dimethylaminopyridin, dimethylaminopyridine, 2-dimethylamino pyridine, 2-dimethylaminopyridine cobalt complex, 2-pyridinamine, n,n-dimethyl, dimethylamino-2 pyridine, dimethylaminopyridin, unii-18q95u3z8h, pyridine, 2-dimethylamino
PubChem CID 21885
IUPAC Name N,N-dimethylpyridin-2-amine
SMILES CN(C)C1=CC=CC=N1

Specifications

Density 1.008
Boiling Point 190°C to 192°C
Flash Point 80°C (176°F)
Refractive Index 1.559
Quantity 5 g
Beilstein 110353
Solubility Information Soluble in water.
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 2-(Dimethylamino)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.