missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)guanosine-d6, TRC

Product Code. 30381567
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381567 5 mg 5mg
30378592 1 mg -
30373517 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381567 Supplier Toronto Research Chemicals Supplier No. TRCD4608525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12H11D6N5O5
Molecular Weight (g/mol) 317.33
Synonym N,N-Dimethylguanosine-d6, 2,2-Dimethylguanosine-d6, 2-Dimethylamino-6-oxypurine Riboside-d6, N2,N2-Dimethylguanosine-d6
SMILES O[C@H]1[C@@H](O)[C@H](N2C(N=C(N(C([2H])([2H])[2H])C([2H])([2H])[2H])NC3=O)=C3N=C2)O[C@@H]1CO

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H17N5O5/c1-16(2)12-14-9-6(10(21)15-12)13-4-17(9)11-8(20)7(19)5(3-18)22-11/h4-5,7-8,11,18-20H,3H2,1-2H3,(H,14,15,21)/t5-,7-,8-,11-/m1/s1/i1D3,2D3
Chemical Name or Material 2-(Dimethylamino)guanosine-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.