missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethylhydrazine Dihydrochloride, TRC

Product Code. 30382935
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382935 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382935 Supplier Toronto Research Chemicals Supplier No. TRCD471530250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 57659-80-0
Molecular Formula C4H13N3 . 2HCl
Quantity 250 mg
Synonym 2-Hydrazino-N,N-dimethylethanamine Dihydrochloride, 2-Hydrazino-N,N-dimethyl-ethanamine Dihydrochloride, [2-(Dimethylamino)ethyl]-hydrazine Dihydrochloride, 2-Hydrazinyl-N,N-dimethylethanamine Dihydrochloride, 2-Hydrazino-N,N-dimethylethylamine Dihydrochloride
InChI Formula InChI=1S/C4H13N3.2ClH/c1-7(2)4-3-6-5;;/h6H,3-5H2,1-2H3;2*1H
SMILES Cl.Cl.CN(C)CCNN
Molecular Weight (g/mol) 176.09
Chemical Name or Material 2-(Dimethylamino)ethylhydrazine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.