missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol

Product Code. 11303117
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303117 100 mL 100mL
11393107 25 mL 25mL
11313117 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11303117 Supplier Thermo Scientific Alfa Aesar Supplier No. L13506.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2439-35-2 | C7H13NO2 | 143.19 g/mol

2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.

Solubility
Soluble in water (1.000 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Sensitive to heat, light and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2439-35-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00038233
InChI Key DPBJAVGHACCNRL-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl acrylate, dimethylaminoethyl acrylate, n,n-dimethylaminoethyl acrylate, adame, 2-propenoic acid, 2-dimethylamino ethyl ester, acrylic acid, 2-dimethylamino ethyl ester, 2-dimethylamino, unii-5yes121ftl, 2-n,n-dimethylamino ethyl acrylate, ageflex fa-1
PubChem CID 17111
IUPAC Name 2-(dimethylamino)ethyl prop-2-enoate
SMILES CN(C)CCOC(=O)C=C

Specifications

Melting Point -60°C
Density 0.943
Boiling Point 64°C (12 mmHg)
Flash Point 62°C (144°F)
Linear Formula CH2=CHCO2CH2CH2N(CH3)2
Refractive Index 1.438
Quantity 100 mL
UN Number UN3302
Beilstein 1099119
Sensitivity Light sensitive
Solubility Information Soluble in water (1.000g/L at 20°C).
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material 2-(Dimethylamino)ethyl acrylate, stabilized with ≈0.1% 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.