missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)-3-quinolinecarbaldehyde, 97+%, Thermo Scientific™

Product Code. 10209222
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10209222 1 g 1g
10299212 250 mg 250mg
10507474 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10209222 Supplier Thermo Scientific Maybridge Supplier No. MO07114DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 728035-61-8
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD06010203
InChI Key RCRLBHNGTPRGJQ-UHFFFAOYSA-N
PubChem CID 1076116
IUPAC Name 2-(dimethylamino)quinoline-3-carbaldehyde
SMILES CN(C)C1=C(C=O)C=C2C=CC=CC2=N1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-dimethylamino quinoline-3-carbaldehyde, 2-dimethylamino-quinoline-3-carbaldehyde, 2-dimethylamino-3-quinolinecarbaldehyde, 3-quinolinecarboxaldehyde,2-dimethylamino, 2-dimethylamino-3-quinoline carbaldehyde, 2-dimethylamino quinoline-3-carboxaldehyde, 3-quinolinecarboxaldehyde, 2-dimethylamino
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material 2-(Dimethylamino)-3-quinolinecarbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.