missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethoxyphosphoryl)acetic acid, TRC

Product Code. 30380524
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380524 250 mg
30386879 500 mg
30384754 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380524 Supplier Toronto Research Chemicals Supplier No. TRCD479333250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34159-46-1
Molecular Formula C4H9O5P
Molecular Weight (g/mol) 168.09
SMILES COP(=O)(CC(=O)O)OC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C4H9O5P/c1-8-10(7,9-2)3-4(5)6/h3H2,1-2H3,(H,5,6)
Formula Weight 168.0188
Chemical Name or Material 2-(Dimethoxyphosphoryl)acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.