missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Diisopropylamino)ethanol, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 2-(Diisopropylamino)ethanol |
| CAS | 96-80-0 |
| Molecular Formula | C8H19NO |
| Quantity | 500 mg |
| Synonym | 2-(Diisopropylamino)ethyl Alcohol, 2-(N,N-Diisopropylamino)ethanol, 2-[Bis(propan-2-yl)amino]ethan-1-ol, DIPAE, Diisopropylethanolamine, Monoethanoldiisopropylamine, N,N-Diisopropyl-2-aminoethanol, N,N-Diisopropylethanolamine, NSC 45475, 2-[Bis(1-methylethyl)amino]ethanol(Diisopropylamino)ethanol, (N,N-Diisopropylamino)ethanol |
| InChI Formula | InChI=1S/C8H19NO/c1-7(2)9(5-6-10)8(3)4/h7-8,10H,5-6H2,1-4H3 |
| SMILES | CC(C)N(CCO)C(C)C |
| IUPAC Name | 2-[di(propan-2-yl)amino]ethanol |
| Molecular Weight (g/mol) | 145.24 |
| Formula Weight | 145.1467 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction