missing translation for 'onlineSavingsMsg'
Learn More

2-(Difluoromethoxy)benzylamine, 97%, Thermo Scientific Chemicals

Product Code. 11384046
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384046 1 g 1g
11394046 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384046 Supplier Thermo Scientific Alfa Aesar Supplier No. L19165.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 243863-36-7 | Molecular Formula: C8H9F2NO | Molecular Weight: 173.163 g/mol

2-(Difluoromethoxy)benzylamine is used as a pharmaceutical intermediate. It is also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Difluoromethoxy)benzylamine is used as a pharmaceutical intermediate. It is also used in chemical research.

Notes
Incompatible with oxidizing agents, water and bases.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 243863-36-7
Molecular Formula C8H9F2NO
Molecular Weight (g/mol) 173.163
MDL Number MFCD00236229
InChI Key GGXXUJUQXAOYPK-UHFFFAOYSA-N
Synonym 2-difluoromethoxy benzylamine, 2-difluoromethoxy phenyl methanamine, 1-2-difluoromethoxy phenyl methanamine, benzenemethanamine, 2-difluoromethoxy, 2-difluoromethoxy phenyl methylamine, acmc-20a5tk, 2-difluoromethoxy-benzylamine, 2-difluoromethoxy benzyl amine
PubChem CID 1514062
IUPAC Name [2-(difluoromethoxy)phenyl]methanamine
SMILES C1=CC=C(C(=C1)CN)OC(F)F

Specifications

Refractive Index 1.491
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 173.16
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethoxy)benzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.