missing translation for 'onlineSavingsMsg'
Learn More

2-(Difluoromethoxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11389839
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11389839

Brand: Thermo Scientific Alfa Aesar B24272.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-(Difluoromethoxy)benzaldehyde is an important raw material used as an intermediate in the preparation of pharmaceuticals, agrochemicals, and dyestuffs. It is also involved in the preparation of 4-(2-difluoromethoxyphenyl)- substituted 1,4-dihydropyridine 3,5-diesters.

Solubility
Miscible with 2-propanol and acetic acid.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71653-64-0
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00042251
InChI Key QPBNHDFPMRENBC-UHFFFAOYSA-N
Synonym 2-difluoromethoxy benzaldehyde, 2-difluoromethoxy-benzaldehyde, o-difluoromethoxybenzaldehyde, 2-difloromethoxybenzaldehyde, pubchem2906, acmc-209ojk, difluoromethoxybenzaldehyde, ksc356m9l, zerenex zx009675, benzaldehyde, difluoromethoxy
PubChem CID 2736984
IUPAC Name 2-(difluoromethoxy)benzaldehyde
SMILES C1=CC=C(C(=C1)C=O)OC(F)F

Specifications

Density 1.284
Boiling Point 88°C to 90°C (9 mmHg)
Flash Point 106°C (223°F)
Refractive Index 1.497
Quantity 1 g
Beilstein 389068
Sensitivity Air Sensitive
Solubility Information Miscible with 2-propanol and acetic acid.
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethoxy)benzaldehyde
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.