missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)-N-(2,5-dimethylphenyl)acetamide Hydrochloride, TRC

Product Code. 30011656
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011656 250 mg 250mg
30138462 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011656 Supplier Toronto Research Chemicals Supplier No. TRCD443595250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1012864-23-1
Molecular Formula C14 H22 N2 O . Cl H
Molecular Weight (g/mol) 270.8
Synonym 2-(Diethylamino)-N-(2,5-dimethylphenyl)acetamide Hydrochloride, Lidocaine Hydrochloride Imp. J (EP) as Hydrochloride
IUPAC Name 2-(diethylamino)-N-(2,5-dimethylphenyl)acetamide;hydrochloride
SMILES Cl.CCN(CC)CC(=O)Nc1cc(C)ccc1C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H22N2O.ClH/c1-5-16(6-2)10-14(17)15-13-9-11(3)7-8-12(13)4;/h7-9H,5-6,10H2,1-4H3,(H,15,17);1H
Formula Weight 270.1499
Percent Purity >95%
Chemical Name or Material 2-(Diethylamino)-N-(2,5-dimethylphenyl)acetamide Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.