missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)-N-(2,5-dimethylphenyl)acetamide Hydrochloride, Mikromol™

Product Code. 30049753
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30049753

Brand: Mikromol MM0102.130025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(Diethylamino)-N-(2,5-dimethylphenyl)acetamide Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Diethylamino)-N-(2,5-dimethylphenyl)acetamide Hydrochloride
CAS 1012864-23-1
Analyte or Component Names 2-(Diethylamino)-N-(2,5-dimethylphenyl)acetamide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H22 N2 O . Cl H
Certifications/Compliance ISO 17025
Synonym 2-(Diethylamino)-N-(2,5-dimethylphenyl)acetamide Hydrochloride, Lidocaine Hydrochloride Imp. J (EP) as Hydrochloride, Lidocaine Hydrochloride Imp. J (EP), Lidocaine Imp. J (EP), Lidocaine Hydrochloride Impurity J as Hydrochloride, Lidocaine Impurity J as Hydrochloride
InChI Formula InChI=1S/C14H22N2O.ClH/c1-5-16(6-2)10-14(17)15-13-9-11(3)7-8-12(13)4;/h7-9H,5-6,10H2,1-4H3,(H,15,17);1H
SMILES Cl.CCN(CC)CC(=O)Nc1cc(C)ccc1C
IUPAC Name 2-(diethylamino)-N-(2,5-dimethylphenyl)acetamide;hydrochloride
Molecular Weight (g/mol) 270.8
Quantity 25 mg
Formula Weight 270.1499 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.