missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)-N-(2,3-dimethylphenyl)acetamide Hydrochloride, TRC

Product Code. 30136540
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136540 2.5 g 2.5g
30007737 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136540 Supplier Toronto Research Chemicals Supplier No. TRCD4437452.5G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 857170-72-0
Molecular Formula C14 H22 N2 O . Cl H
Molecular Weight (g/mol) 270.8
Synonym 2-(Diethylamino)-N-(2,3-dimethylphenyl)acetamide hydrochloride, Lidocaine Hydrochloride Imp. F (EP) as Hydrochloride
IUPAC Name 2-(diethylamino)-N-(2,3-dimethylphenyl)acetamide;hydrochloride
SMILES Cl.CCN(CC)CC(=O)Nc1cccc(C)c1C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H22N2O.ClH/c1-5-16(6-2)10-14(17)15-13-9-7-8-11(3)12(13)4;/h7-9H,5-6,10H2,1-4H3,(H,15,17);1H
Formula Weight 270.1499
Percent Purity >95%
Chemical Name or Material 2-(Diethyl-amino)-N-(2,3-dimethylphenyl)acetamide Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.